ReadiLink™ Rapid iFluor® 350 Antibody Labelling Kit
Supplier: AAT BIOQUEST INC
5700
77462-906EA 523.82 USD
77462-906
ReadiLink™ Rapid iFluor® 350 Antibody Labelling Kit
Protein Linking and Labeling Systems
ReadiLink™ Rapid antibody labelling kits, designed for production scale, provide a convenient and efficient method for labelling large volumes of antibodies with our superior iFluor® dyes.
  • Robust protocol
  • Consistent results
  • Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
Delivery information: Shipped at RT.
Caution: Research use only.