Buccutite™ Rapid PerCP Antibody Labelling Kit
Supplier: AAT Bioquest
5407
77462-880EA 1501.75 USD
77462-880
Buccutite™ Rapid PerCP Antibody Labelling Kit
Protein Linking and Labeling Systems
Production scale optimized for labelling 1 mg antibody per reaction. Buccutite™ Rapid PerCP antibody labelling kits, designed for large-scale production, provide a streamlined approach for labelling antibodies with PerCP, APC, PE and iFluor® tandem dyes.
  • Robust protocol
  • Consistent results
  • Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
Delivery information: Shipped with blue ice.
Caution: Research use only.