mFluor™ Violet 545 styramide
Supplier: AAT Bioquest
45078
77462-296EA 318.85 USD
77462-296
mFluor™ Violet 545 styramide
mFluor™ Violet 545 styramide
mFluor™ Violet 545 Styramide uses the bright green fluorescent dye mFluor™ Violet 545 (Ex/Em = 393/543 nm) to label targets in situ.
  • Stable for 12 months
The Power Styramide™ Signal Amplification (PSA™) system is a highly sensitive method for detecting low-abundant targets in fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC), and in situ hybridization (ISH).
Caution: For Research Use Only