mFluor™ Red 780 styramide
Supplier: AAT Bioquest
45076
77462-292EA 318.85 USD
77462-292
mFluor™ Red 780 styramide
mFluor™ Red 780 styramide
mFluor™ Red 780 Styramide uses the bright red fluorescent dye mFluor™ Red 780 (Ex/Em = 629/767 nm) to label targets in situ.
  • Stable for 12 months
The Power Styramide™ Signal Amplification (PSA™) system is a highly sensitive method for detecting low-abundant targets in fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC), and in situ hybridization (ISH).
Caution: For Research Use Only