mFluor™ Blue 630 styramide
Supplier: AAT Bioquest
45073
77462-286EA 318.85 USD
77462-286
mFluor™ Blue 630 styramide
mFluor™ Blue 630 styramide
The Power Styramide™ Signal Amplification (PSA™) system is a highly sensitive method for detecting low-abundant targets in fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC), and in situ hybridization (ISH).
  • Stable for 12 months
Quality tested in-house
Caution: For Research Use Only