iFluor® 660 styramide
Supplier: AAT Bioquest
44906
77462-268EA 327.22 USD
77462-268
iFluor® 660 styramide
iFluor® 660 dyes have fluorescence excitation and emission maxima of ~660 nm and ~680 nm respectively.
  • Stable for 12 months
Power Styramide™ Signal Amplification (PSA™) system is one of the most sensitive methods that can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10-50 times higher than the widely used tyramide (TSA) reagents.
Caution: For Research Use Only