Amplite® Tyramide (TSA) Signal Booster
Supplier: AAT BIOQUEST INC
44502
77462-258EA 100.94 USD
77462-258
Amplite® Tyramide (TSA) Signal Booster
Nucleic Acid Reagents Nucleic Acid Modification and Labelling
Tyramide signal amplification (TSA) is a technique used to enhance the signal of target molecules, such as antibodies or nucleic acid probes, in immunohistochemistry and in situ hybridization.
  • Stable for 12 months
Quality tested in-house.
Caution: For research use only.