Psoralen MOP azide
Supplier: AAT BIOQUEST INC
39057
77462-242EA 202.92 USD
77462-242
Psoralen MOP azide
Psoralen MOP azide
Psoralen MOP Azide is an excellent building block for developing site-specific nucleic acid probes via the well-known click chemistry, i.e., Copper-Catalyzed Alkyne–Azide Cycloaddition (CuAAC).
  • Stable for 24 months
Quality tested in-house
Caution: For Research Use Only