mFluor™ Violet 540 Biotin conjugate
Supplier: AAT Bioquest
3114
77462-188EA 235.39 USD
77462-188
mFluor™ Violet 540 Biotin conjugate
mFluor™ Violet 540 Biotin
mFluor™ Violet 540 Biotin Conjugate contains mFluor™ Violet 540 fluorophore that is well excited by the common 405 nm violet laser.
  • Stable for 12 months
Quality tested in-house
Caution: For Research Use Only