ReadiUse™ Universal Kinase Reaction Stopping Buffer
Supplier: AAT BIOQUEST INC
31005
77462-178EA 100.94 USD
77462-178
ReadiUse™ Universal Kinase Reaction Stopping Buffer
Buffers Cloning Buffers
ReadiUse™ Universal Kinase Reaction Stopping Buffer is an aqueous solution that effectively chelates divalent metal ions.
  • Designed for ease of use
  • Saves time
This sterile-filtered solution finds wide applicability in biochemistry and molecular biology research.
Caution: For research use only.