ATTO 647N acid
Supplier: AAT BIOQUEST INC
2854
77462-150EA 591.05 USD
77462-150
ATTO 647N acid
ATTO 647N acid
ATTO 647N is a rhodamine-based fluorescent dye optimized for use in the red spectral region, with similar spectral characteristics as Cy5.
  • Stable for 12 months
Quality tested in-house
Caution: For Research Use Only