mFluor™ Violet 500-Wheat Germ Agglutinin (WGA) Conjugate
Supplier: AAT BIOQUEST INC
25452
77462-012EA 152.71 USD
77462-012
mFluor™ Violet 500-Wheat Germ Agglutinin (WGA) Conjugate
Protein Linking and Labeling Systems
mFluor™ Violet 500 labeled WGA is well-excited by the violet laser, emitting a bright green fluorescence at 501 nm.
  • Stable for 12 months
Wheat germ agglutinin (WGA) is a well-studied lectin known for its binding affinity to N-acetyl-D-glucosamine and sialic acid, making it a valuable tool in various biological applications.
Caution: For research use only.