ReadiUse™ Yeast Cell Lysis Buffer, 5X
Supplier: AAT BIOQUEST INC
24102
77461-950EA 116.17 USD
77461-950
ReadiUse™ Yeast Cell Lysis Buffer, 5X
Buffers Extraction Buffers
ReadiUse™ Yeast Cell Lysis Buffer is a biochemical reagent designed for efficient yeast cell lysis and protein solubilization in laboratory applications.
  • Designed for ease of use
  • Saves time
  • Versatile in its efficacy across various organisms, including Bacillus subtilis, Gram-positive, and Gram-negative bacteria
With a mild detergent formulation, ReadiUse™ Yeast cell lysis buffer minimizes the reliance on harsh chemical treatments, extreme pH adjustments, or temperature variations, contributing to the preservation of extracted protein integrity.
Caution: For research use only.