LysoBrite™ Green DND-26
Supplier: AAT BIOQUEST INC
22648
77461-914EA 157.01 USD
77461-914
LysoBrite™ Green DND-26
LysoBrite™ Green DND-26
LysoBrite™ Green DND-26 is chemically the same as the LysoTracker® Green DND-26 used for labeling and tracking acidic organelles in live cells (LysoTracker® is the trademark of ThermoFisher).
  • Stable for 6 months
Quality tested in-house
Caution: For Research Use Only