MESG phosphate assay reagent

Supplier: AAT BIOQUEST INC

21600-1MG
77461-882EA 81.42 USD
77461-882
MESG phosphate assay reagent
MESG

In the presence of inorganic phosphate MESG is converted to 2-amino-6-mercapto-7-methlpurine by purine nucleoside phosphorylase (EC 2.4.2.1) with absorption wavelength shift to red.


  • Stable for 12 months


Quality tested in-house

MW: 313.34 g/mol
CAS Number: 55727-10-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR