7.2WS maleimide [TQ7.2WS maleimide], Tide Quencher™
Supplier: AAT BIOQUEST INC
2126
77461-846EA 235.39 USD
77461-846
7.2WS maleimide [TQ7.2WS maleimide], Tide Quencher™
7.2WS maleimide [TQ7.2WS maleimide]
Tide Quencher™ 7.2WS (TQ7.2WS) is a non-fluorescent molecule designed to efficiently quench the fluorescence of common NIR fluorophores such as Cy7, Alexa Fluor® 700, Alexa Fluor® 750, iFluor® 700, iFluor® 710, iFluor® 720 and iFluor® 750.
  • Stable for 12 months
  • Optimized for FRET applications
Quality tested in-house
Caution: For Research Use Only