Gelite™ 100 bp to 1 kb DNA Ladder
Supplier: AAT BIOQUEST INC
17760
77461-676EA 100.94 USD
77461-676
Gelite™ 100 bp to 1 kb DNA Ladder
Electrophoresis Reagents Molecular Weight Markers
The Gelite™ 100 bp to 1 kb DNA ladder is designed to help size and estimate the mass of double-stranded DNA and PCR products within the 100 to 10000 base pair range on agarose or polyacrylamide gels.
  • Stable for 12 months
Quality tested in-house.
Caution: For research use only.