Gelite™ 100 bp DNA Ladder
Supplier: AAT BIOQUEST INC
17753
77461-672EA 100.94 USD
77461-672
Gelite™ 100 bp DNA Ladder
Electrophoresis Reagents Molecular Weight Markers
The Gelite™ 100 bp DNA Ladder is designed for sizing and estimating the mass of double-stranded DNA and PCR products within the 100 to 1500 bp range on agarose or polyacrylamide gels.
  • Stable for 12 months
Quality tested in-house.
Caution: For research use only.