Gelite™ Safe DNA Gel Stain *GelRed Alternative, 10000X in DMSO*
Supplier: AAT BIOQUEST INC
17711
77461-668EA 106.26 USD
77461-668
Gelite™ Safe DNA Gel Stain *GelRed Alternative, 10000X in DMSO*
Stains Electrophoresis Stains
Gelite™ Safe has been developed specifically to be less hazardous than EtBr for staining DNA in agarose and acrylamide gels with much higher sensitivity.
  • Stable for 12 months
AAT Bioquest is committed to designing our products to be environment-friendly.
Caution: For research use only.