Propidium iodide 1 mg/ml in aqueous solution

Supplier: AAT Bioquest

17585
77461-644EA 112.31 USD
77461-644
Propidium iodide 1 mg/ml in aqueous solution

Propidium iodide (PI) is a red-fluorescent dye widely used for staining and identifying dead cell populations in flow cytometry and fluorescence microscopy.


  • Stable for 6 months


PCR (polymerase chain reaction) is a laboratory technique used to amplify DNA (deoxyribonucleic acid) segments.


Caution: For Research Use Only

Formula: C₂₇H₃₄I₂N₄
MDL Number: MFCD00011921
CAS Number: 25535-16-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR