ReadiUse™ dNTP Mix Set *10 mM PCR Grade*
Supplier: AAT BIOQUEST INC
17258
77461-568EA 84.71 USD
77461-568
ReadiUse™ dNTP Mix Set *10 mM PCR Grade*
Nucleic Acid Reagents Nucleotides
10 mM dNTP Mix is a mixture of four nucleotides (dATP, dCTP, dGTP, dTTP) in purified water.
  • Each nucleotide is at a concentration of 10 mM
  • The formulation is stable for 12 months at −20 °C
10 mM dNTP mix is suitable for use in polymerase chain reaction (PCR), sequencing, fill-in reactions, nick translation, cDNA synthesis, and TdT-tailing reactions.
Caution: For research use only.