dTTP *100 mM PCR Grade*
Supplier: AAT BIOQUEST INC
17256 17257
77461-564EA 106.26 USD
77461-564 77461-566
dTTP *100 mM PCR Grade*
Nucleic Acid Reagents Nucleotides
2'-Deoxythymidine-5'-Triphosphate (dTTP) solution is used in DNA synthesis reactions such as PCR, DNA sequencing and molecular cloning techniques.
  • Highly purified
DNA sequencing is a fundamental tool in biological and medical research (especially for personalized medicine).
Caution: For research use only.