iFluor® 488-dUTP *1 mM in TE Buffer (pH 7.5)*
Supplier: AAT BIOQUEST INC
17044
77461-530EA 333.31 USD
77461-530
iFluor® 488-dUTP *1 mM in TE Buffer (pH 7.5)*
Nucleic Acid Reagents Nucleotides
This iFluor 488-dUTP conjugate is used as a green fluorescence color with SpectrumGreen™ filter set (SpectrumGreen™ is the trademark of Vysis).
  • Widely used for both FISH probes and microarray-based experiments
  • Bright signal that has high photostability and is not affected by pH
The dye-modified deoxyuridine 5'-triphosphates are one of the most common methods to produce dye-labelled DNA via the conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labeling or PCR.
Caution: For research use only.