Fluorescein-12-dGTP *1 mM in Tris Buffer (pH 7.5)*
Supplier: AAT BIOQUEST INC
17037
77461-522EA 106.26 USD
77461-522
Fluorescein-12-dGTP *1 mM in Tris Buffer (pH 7.5)*
Nucleic Acid Reagents Nucleotides
Fluorescein-12-dGTP might be incorporated into a DNA sequence as its natural counterpart dGTP to incorporate a fluorescein tag in a DNA molecule.
  • Can be used for the direct enzymatic labeling of DNA/cDNA, e.g., by PCR or nick translation
The linker between fluorescein tag and guanosine has been optimized for the optimal substrate incorporation and labeling efficiency.
Caution: For research use only.