iFluor® 625 succinimidyl ester
Supplier: AAT Bioquest
1422
77461-476EA 235.39 USD
77461-476
iFluor® 625 succinimidyl ester
iFluor® 625 succinimidyl ester
iFluor® 625 succinimidyl ester is the most convenient reactive form of iFluor® 625 dyes that can be readily used for labeling proteins such as antibodies.
  • Stable for 12 months
Quality tested in-house
Caution: For Research Use Only