iFluor® 780 succinimidyl ester
Supplier: AAT BIOQUEST INC
1371
77461-460EA 235.39 USD
77461-460
iFluor® 780 succinimidyl ester
iFluor® 780 succinimidyl ester
iFluor® 780 dyes have fluorescence excitation and emission maxima of ~784 nm and ~808 nm respectively.
  • Stable for 12 months
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, particularly antibodies.
Caution: For Research Use Only