Buccutite™ Rapid APC-iFluor® 750 Tandem Antibody Labelling Kit
Supplier: AAT BIOQUEST INC
1348
77461-454EA
353.61
USD
77461-454
Buccutite™ Rapid APC-iFluor® 750 Tandem Antibody Labelling Kit
Protein Linking and Labeling Systems
Microscale optimized for labelling 25 µg antibody per reaction. APC-iFluor® 750 tandem is an excellent replacement for APC-Cy7 and APC-Alexa Fluor® 750 tandem since they have almost identical spectra.
- Robust protocol
- Consistent results
- Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
Delivery information: Shipped with blue ice.
Caution: Research use only.