Ac-DEVD-aminoluciferin
Supplier: AAT Bioquest
13210
77461-436EA 100.94 USD
77461-436
Ac-DEVD-aminoluciferin
Ac-DEVD-aminoluciferin
Ac-DEVD-aminoluciferin is a luminogenic substrate that measures caspase-3 and -7 activities in apoptotic cells.
  • Stable for 24 months
Quality tested in-house
Caution: For Research Use Only