Buccutite™ Rapid trFluor™ D2 Acceptor Antibody Labelling Kit
Supplier: AAT BIOQUEST INC
1302
77461-428EA 1320.37 USD
77461-428
Buccutite™ Rapid trFluor™ D2 Acceptor Antibody Labelling Kit
Protein Linking and Labeling Systems
Microscale optimized for labelling 100 µg antibody per reaction. Many biological compounds present in cells, serum, or other biological fluids are naturally fluorescent, and thus the use of conventional, prompt fluorophores leads to serious limitations in assay sensitivity due to the high background caused by the autofluorescence of the biological molecules to be assayed.
  • Robust protocol
  • Consistent results
  • 12-month shelf life
Designed to provide high quality labelled antibodies for multiple downstream applications.
Delivery information: Shipped at RT.
Caution: Research use only.