iFluor® 690 succinimidyl ester
Supplier: AAT Bioquest
1042
77461-352EA 327.22 USD
77461-352
iFluor® 690 succinimidyl ester
iFluor® 690 succinimidyl ester
iFluor® 690 dyes have fluorescence excitation and emission maxima of ~685 nm and ~704 nm respectively.
  • Stable for 12 months
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, particularly antibodies.
Caution: For Research Use Only