PBS Tablet (Phosphate Buffered Saline Tablet)
Supplier: AAT BIOQUEST INC
10010
77461-346EA 47.93 USD
77461-346
PBS Tablet (Phosphate Buffered Saline Tablet)
Buffers Electrophoresis Buffers Washing Buffers
Each PBS tablet prepares 100 mL of a 1X PBS solution containing 137 mM sodium chloride, 10 mM phosphate buffer, and 2.7 mM potassium chloride (pH 7.4).
  • Designed for ease of use
  • Saves time
PBS (phosphate buffered saline) is an isotonic buffer that closely mimics the human body's pH, osmolarity, and ion concentrations.
Caution: For research use only.