BioXp® Select mRNA Synthesis Kits
Supplier: TELESIS BIO
The BioXp Select mRNA synthesis kit enables the synthesis of mRNA from an input DNA template.
- mRNA size: 0.4 to 10 kb
- Yield: 50 µg/well
- Compatible with BioXp® 9600
- Cap: 5' CleanCap® AG (3'OMe)
- Modification: Pseudouridine
The BioXp® Select mRNA synthesis kit enables you to synthesize purified capped and tailed mRNA between 0.4 to 10 kb. The kit contains all the necessary reagents for on-demand synthesis of mRNA using a linear DNA template generated with a plasmid or PCR product for transcription.