BioXp® 3250 Systems

Supplier: TELESIS BIO
BX3250-01
77461-216EA 134330 USD
77461-216
BioXp® 3250 Systems
Next Generation Sequencing Instruments
The next generation high-throughput synthetic biology workstation for accelerating discovery.

  • Gene synthesis: Custom high-fidelity gene fragments
  • mRNA: Custom
  • Synthetic mRNA
  • DNA cloning: Custom DNA fragments, up to 7,000 bp, cloned into any vector
  • DNA amplification: Cell-free DNA amplification
  • Libraries: Custom variant DNA libraries

The BioXp™ system has empowered researchers worldwide to improve productivity, reduce turnaround time, and have complete workflow control. The solution enables synthesis of DNA, mRNA and variant libraries from digital sequence in just days and requires no prior knowledge or expertise to operate.

Researchers can securely submit sequences through the myBioXperience™ portal without risking any transfer of proprietary vector information and build sequences in their own lab using push-button automation on their own schedule.

Certifications: NRTL.

Accessories information: Power input voltage: 100 to 240 V
Power input current: 8.3 A Max
Operating temperature range: 16...40 °C
Weight: 63.4 kg (139.8 US lbs)
Dimensions (W×D×H): 69×77×53 cm (27×30×21)"

Delivery information: Expected shipping: 40 days.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR