VWR® REDISHIP Polyurethane Laboratory Chairs
Supplier: VWR International
77449-176 77449-178 77449-180 77449-182 77449-184
77449-176EA 342.89 USD
77449-176 77449-178 77449-180 77449-182 77449-184
VWR® REDISHIP Polyurethane Laboratory Chairs
Chairs
VWR® polyurethane seating is perfect for use in almost any environment with its enhanced durability and functionality.
  • Rugged and durable
  • Chemical-resistant
  • Non-porous
  • Easy to clean
  • Convenient adjustability
  • Ships assembled (requires only backbar insertion)
  • Equipped with both glides and casters
  • Weight capacity: 300 lbs.
  • American made
  • 15 year warranty
VWR® polyurethane seating features ergonomic adjustments for seat height and locking back tilt, a nylon-reinforced fiberglass base shipped with both dual-wheel, free-rolling casters and stationary glides.More Product Information