Human Recombinant respiratory syncytial virus (RSV) (B, strain 18537) Nucleoprotein Protein (His Tag) (From E. coli)
Supplier: Sino Biological
40822-V07E-100
77447-826EA 498 USD
77447-826
Human Recombinant respiratory syncytial virus (RSV) (B, strain 18537) Nucleoprotein Protein (His Tag) (From E. coli)
Proteins and Peptides
A DNA sequence encoding the Human respiratory syncytial virus (RSV) (B, strain 18537) Nucleoprotein (P24566) (Met1-Leu391) was expressed with a polyhistidine tag at the N-terminus.