SARS-CoV-2 Recombinant Spike S1+S2 (Q52R, A67V, HV69-70 deletion, Y144 deletion, E484K, D614G, Q677H, F888L) Protein (ECD, His Tag) (from Baculovirus-Insect Cells)
Supplier: Sino Biological
40589-V08B20-100
77447-756EA 448 USD
77447-756
SARS-CoV-2 Recombinant Spike S1+S2 (Q52R, A67V, HV69-70 deletion, Y144 deletion, E484K, D614G, Q677H, F888L) Protein (ECD, His Tag) (from Baculovirus-Insect Cells)
Proteins and Peptides
A DNA sequence encoding the SARS-CoV-2 Spike S1+S2 (YP_009724390.1, with mutations Q52R, A67V, HV69-70 deletion, Y144 deletion, E484K, D614G, Q677H, F888L) (Met1-Pro1213) was expressed with a polyhistidine tag at the C-terminus. The mutations were identified in the SARS-CoV-2 variant (known as variant B.1.525) which emerged in the Multiple countries.