PTS Connect® CardioChek Printer Labels Roll
Supplier: PTS Diagnostics
PTS2151
77445-750EA 15.85 USD
77445-750
PTS Connect® CardioChek Printer Labels Roll
Labels
For best results, the use of PTS Connect printer labels is recommended.
  • Labels are sensitive to heat, moisture, and light
  • Store labels at room temperature, in a cool, dry location
  • Do not expose labels to high heat or direct light for an extended period of time
  • Not for use with organic solvents
  • One roll of 160 labels
The PTS Connect printer label roll is designed to be used with the PTS Connect printer. The printer labels have a sensor mark on the backing of each label for top-of-label alignment. Check to be sure the labels have this mark before loading them into the printer.