Phosphate Buffered Saline (PBS) 1X with 0.05% Tween 20, pH 7.4
Supplier: TEKNOVA INC
P0276
77442-720EA 535.46 USD
77442-720
Phosphate Buffered Saline (PBS) 1X with 0.05% Tween 20, pH 7.4
Buffers Cell Culture Buffers
1X Solution composition: 137 mM Sodium chloride, 2.7 mM potassium chloride, 4.3 mM sodium phosphate, 1.4 mM potassium phosphate and 0.05% Tween (v/v).