PHCbi High Performance Undercounter Lab Refrigerator
Supplier: PHC Corporation
TSU-4RW-N6
77442-056EA 4004.55 USD
77442-056
PHCbi High Performance Undercounter Lab Refrigerator
Refrigerators
The PHCbi brand TSU-4RW-N6 is developed for the critical storage of vaccines and biologics, and is designed to meet current and evolving CDC, NSF, and ANSI guidelines. This ADA compliant undercounter is Energy Star Certified and combines high performance temperature control with environmentally friendly refrigerants in a compact, glass door design.
  • Temperature control and ±3 °C uniformity
  • Security and alarm notification
  • Self closing glass door
  • SNAP compliant refrigerant
  • ADA compliant design
  • Optional stacking kit
The TSU-4RW-N6 is an energy efficient ADA compliant high performance undercounter Refrigerator engineered to meet the strict regulatory guidelines. This model features a microprocessor controller, digital display and intuitive operation in a small footprint. This unit ensures precise temperature control and uniformity through a refrigeration system utilizing hydrocarbon refrigerants. This model includes an access port for an external monitoring system setup.
Temperature sensor: Thermistor
Certifications: Energy Star Certified; Meets CDC/VFC, NSF/ANSI-456 regulatory guidelines; SNAP compliant refrigerants.
Ordering information: Optional stacking kit and blackout panel available to order separately.