QuantiFluo™ Adenosine Deaminase Assay Kit
Supplier: BIOASSAY SYSTEMS
DADA-100
77441-646EA 436.6 USD
77441-646
QuantiFluo™ Adenosine Deaminase Assay Kit
Assays Enzyme Assays
For quantitative determination of adenosine deaminase enzyme activity in biological samples.
  • Fast and safe
  • Assay can be completed within 50 minutes
  • Nonradioactive assay
  • Sensitive and accurate
  • Linear detection range is 0.3 to 30 U/L
  • adenosine deaminase in a 96-well plate assay
  • Convenient and high-throughput
  • Homogeneous 'mix-incubatemeasure' type assay
  • Can be readily automated to assay thousands of samples per day
Adenosine deaminase (EC 3.5.4.4) is a key enzyme in purine metabolism and plays an important role in the normal immune function of humans. ADA deficiency is one cause of Severe Combined Immunodeficiency (SCID), and elevated levels of ADA have been observed in patients with tuberculosis and immune-related diseases.
BioAssay systems DADA-100 Kit provides a convenient fluorimetric method to measure adenosine deaminase activity in biological samples. In this assay, liberated ammonia reacts with the reagents where the increase in fluorescence at λex/em = 415/475 nm is directly proportional to enzyme activity.