NEBNext® Primers for Epigenetics (Unique Dual Index Set 2B)
Supplier: New England Biolabs (NEB)
E3392S
77441-300EA
118
USD
77441-300
NEBNext® Primers for Epigenetics (Unique Dual Index Set 2B)
Nucleic Acid Reagents
Next Generation Sequencing Reagents
The NEBNext® Primers for Epigenetics contain unique dual index primers optimized for E5hmC-seq libraries.
- Optimized for NEBNext Epigenetics workflows
This 24-reaction kit is supplied in a plate with premixed i5 and i7 8-base index primers in 24 wells.