Accessories for Compresstome® Machine Oil
Supplier: PRECISIONARY INSTRUMENTS LLC
VFVMOILCANAL
77435-528EA 17.15 USD
77435-528
Accessories for Compresstome® Machine Oil
Microtomes and Microtome Assemblies Microtomes
Machine oil that can be used to lubricate the moving parts on Compresstome models.
  • One 15 ml bottle
Clear machine oil to help maintain the Compresstome tissue slicer. Every few weeks, add a couple of drops to the vibration head unit, then run the machine for five ten 10 minutes continuously to allow the oil to distribute.