Accessories for Compresstome® VF-510-0Z Buffer Tray
Supplier: PRECISIONARY INSTRUMENTS LLC
VFBT510
77435-432EA 331.69 USD
77435-432
Accessories for Compresstome® VF-510-0Z Buffer Tray
Microtomes and Microtome Assemblies Microtomes
A replacement buffer tray for the VF-510-0Z compresstome.
  • Buffer tray made of high-quality, durable Lexan™ polycarbonate
  • Full one year warranty
  • Tube size: 15.5 or 20 mm diameter (must specify)
Packaging: Includes one buffer tray, one set of screws, one gasket, and one adaptor.