Tissue Floating Bath
Supplier: PRECISIONARY INSTRUMENTS LLC
TF1000
77435-368EA 864.53 USD
77435-368
Tissue Floating Bath
Baths Tissue Baths
The precisionary instruments tissue floating bath is a reliable and efficient solution for softening embedded tissue specimens without causing wrinkles or distortions. With a teflon-coated chamber for easy cleaning, a low-profile interior chamber to reduce glare.
  • The tissue floating bath is used after sectioning with a rotary microtome
For use after sectioning paraffin wax.