Rotary Microtome RF-800
Supplier: PRECISIONARY INSTRUMENTS LLC
RF800
77435-338EA 20133.59 USD
77435-338
Rotary Microtome RF-800
Microtomes and Microtome Assemblies Microtomes
Semi-automated rotary microtome for slicing wax embedded fixed samples. The semi-automation comes from the advancement and retraction of the specimen head.
  • Semi-automated rotary microtome
Packaging: Unit includes one RF-800 rotary microtome (with specimen head cassette clamp), one power cord, one removable waste tray, one microtome knife block set, one tissue brush set and forceps, four specimen metal molds, 20 embedding cassettes, and 1 RF-800 user manual.