Rotary Microtome RF-600
Supplier: PRECISIONARY INSTRUMENTS LLC
RF600
77435-314EA 15236.24 USD
77435-314
Rotary Microtome RF-600
Microtomes and Microtome Assemblies Microtomes
Manually operated rotary microtome for slicing wax embedded fixed samples.
  • Manual rotary microtome
Packaging: Unit includes one RF-600 manual rotary microtome (mounted with specimen head cassette clamp), one specimen head paraffin block clamp, one removable waste tray, one microtome knife block set, one high profile blade holder, and one RF-600 user manual.