Laboratory Hot Water Bath
Supplier: PRECISIONARY INSTRUMENTS LLC
HWBATH16L
77435-286EA 935.95 USD
77435-286
Laboratory Hot Water Bath
Baths Water Baths, Unstirred
Laboratory hot water bath with a 16 liters capacity.
  • Use to incubate sections from slicing on a rotary microtome
  • Operating temperature up to 100 °C
  • Digital temperature display and easy controls
Applications include incubation of tissue slices for electrophysiology and precision-cut tissue slices, and paraffin-embedded wax slices from rotary microtome sectioning.