ABS Plastic Indicator for Industial Applications, Defender 3000
Supplier: Ohaus
30659310
77435-132EA 449.54 USD
77435-132
ABS Plastic Indicator for Industial Applications, Defender 3000
Balances and Scales
ABS plastic indicator for basic industrial applications.
  • Results are clearly shown using the large, bright backlit LCD display
  • In Checkweighing mode, the display changes color (yellow/green/red) to alert when it is within the target weight range
Designed with affordability in mind for basic industrial applications, the i-DT33P indicator combines high-impact ABS housing with a large multicolor backlit LCD display and bi-directional RS232. The i-DT33P also features software for Weighing, Parts Counting, Checkweighing, and Totalization.