3-Fluoro-4-hydroxybenzaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

PC9083-100G
77434-086EA 477.82 USD
77434-086
3-Fluoro-4-hydroxybenzaldehyde
3-Fluoro-4-hydroxybenzaldehyde
Formula: C₇H₅FO₂
Melting Pt: 121…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00016626
CAS Number: 405-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR