2',4',5'-Trifluoroacetophenone 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-(2,4,5-Trifluorophenyl)ethan-1-one, 2,4,5-Trifluoroacetophenone

PC7202J-5G
77432-956EA 21.34 USD
77432-956
2',4',5'-Trifluoroacetophenone 99%
2',4',5'-Trifluoroacetophenone
Formula: C₈H₅F₃O
Boiling Pt: 99…101 °C/20 mmHg
Melting Pt: –7 °C
Density: 1.331 g/cm³ (25 °C)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00061193
CAS Number: 129322-83-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR